Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL1R1 SIRNA Assays ›
          Gene Symbol:
          IL1R1
          Gene Name:
          interleukin 1 receptor type 1
          Gene Aliases:
          CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
          Chromosome Location:
          Chr.2: 102069638 - 102179874 on Build GRCh38
          UniGene ID:
          Hs.701982
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 110767

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL1R1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000877.3 NP_000868.1 4 547
          NM_001288706.1 NP_001275635.1 4 460
          NM_001320978.1 NP_001307907.1 4 460
          NM_001320980.1 NP_001307909.1 4 601
          NM_001320981.1 NP_001307910.1 4 442
          NM_001320982.1 NP_001307911.1 4 384
          NM_001320983.1 NP_001307912.1 2 235
          NM_001320984.1 NP_001307913.1 3 302
          NM_001320985.1 NP_001307914.1 3 302
          NM_001320986.1 NP_001307915.1 4 601
          XM_005263930.4 XP_005263987.1 4 409
          XM_005263934.4 XP_005263991.1 5 514
          XM_011511115.2 XP_011509417.1 5 506
          XM_011511116.1 XP_011509418.1 4 484
          XM_011511117.1 XP_011509419.1 4 479
          XM_011511118.2 XP_011509420.1 4 410
          XM_017003989.1 XP_016859478.1 4 607
          GenBank mRNA AK314433.1 - 4 460
          AU136426.1 - 4 588
          BC067506.1 - 3 281 7002107
          BC067507.1 - 3 283 7002109
          BC067508.1 - 3 283 7002111
          BC075062.2 - 3 263 30915370
          BC075063.2 - 3 263 30915586
          BX472731.1 - 4 452
          CD013912.1 - 3 294
          CD013913.1 - 3 294
          CD014140.1 - 2 227
          DA333942.1 - 4 452
          DA861783.1 - 4 460
          M27492.1 - 3 301
          X16896.1 - 4 527

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          cell surface receptor signaling pathway
          response to radiation
          heat acclimation
          ovulation
          regulation of inflammatory response
          interleukin-1-mediated signaling pathway
          response to interleukin-1
          response to epidermal growth factor
          cellular response to glucose stimulus
          response to transforming growth factor beta
          response to nitric oxide
          protease binding
          signal transducer activity
          transmembrane signaling receptor activity
          interleukin-1 receptor activity
          interleukin-1, Type I, activating receptor activity
          platelet-derived growth factor receptor binding
          protein binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0