Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL1R1 SIRNA Assays ›
Gene Symbol:
IL1R1
Gene Name:
interleukin 1 receptor type 1
Gene Aliases:
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
Chromosome Location:
Chr.2: 102069638 - 102179874 on Build GRCh38
UniGene ID:
Hs.701982
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110769
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL1R1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000877.3
NP_000868.1
4
597
NM_001288706.1
NP_001275635.1
4
510
NM_001320978.1
NP_001307907.1
4
510
NM_001320980.1
NP_001307909.1
4
651
NM_001320981.1
NP_001307910.1
4
492
NM_001320982.1
NP_001307911.1
4
434
NM_001320983.1
NP_001307912.1
2
285
NM_001320984.1
NP_001307913.1
3
352
NM_001320985.1
NP_001307914.1
3
352
NM_001320986.1
NP_001307915.1
4
651
XM_005263930.4
XP_005263987.1
4
459
XM_005263934.4
XP_005263991.1
5
564
XM_011511115.2
XP_011509417.1
5
556
XM_011511116.1
XP_011509418.1
4
534
XM_011511117.1
XP_011509419.1
4
529
XM_011511118.2
XP_011509420.1
4
460
XM_017003989.1
XP_016859478.1
4
657
GenBank mRNA
AK314433.1
-
4
510
BC067506.1
-
3
331
7002107
BC067508.1
-
3
333
7002111
BC075062.2
-
3
313
30915370
BC075063.2
-
3
313
30915586
BX472731.1
-
4
502
CD013912.1
-
3
344
CD013913.1
-
3
344
CD014140.1
-
2
277
DA333942.1
-
4
502
DA861783.1
-
4
510
M27492.1
-
3
351
X16896.1
-
4
577
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
immune response
cell surface receptor signaling pathway
response to radiation
heat acclimation
ovulation
regulation of inflammatory response
interleukin-1-mediated signaling pathway
response to interleukin-1
response to epidermal growth factor
cellular response to glucose stimulus
response to transforming growth factor beta
response to nitric oxide
protease binding
signal transducer activity
transmembrane signaling receptor activity
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity
platelet-derived growth factor receptor binding
protein binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0