Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK7 SIRNA Assays ›
          Gene Symbol:
          MAPK7
          Gene Name:
          mitogen-activated protein kinase 7
          Gene Aliases:
          BMK1, ERK4, ERK5, PRKM7
          Chromosome Location:
          Chr.17: 19377721 - 19383544 on Build GRCh38
          UniGene ID:
          Hs.150136
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 110792
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002749.3 NP_002740.2 7 2728
          NM_139032.2 NP_620601.1 5 2536
          NM_139033.2 NP_620602.2 7 2939
          NM_139034.2 NP_620603.2 7 2640
          XM_006721557.3 XP_006721620.1 8 2704
          XM_006721558.3 XP_006721621.1 8 2957
          XM_006721559.3 XP_006721622.1 8 2866
          XM_011523957.2 XP_011522259.1 5 2443
          GenBank mRNA AB209611.1 - 3 4328
          AK295376.1 - 7 2518
          AY534741.1 - 5 2660
          BC007404.2 - 5 2504 3009873
          BC007992.2 - 5 2504 3009873
          BC009963.2 - 7 2660 4300124
          BC030134.1 - 7 2575 4111084
          U25278.1 - 7 2633
          U29725.1 - 7 2774
          U29726.1 - 7 2607

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00177079_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cell cycle
          signal transduction
          peptidyl-serine phosphorylation
          cAMP-mediated signaling
          cell differentiation
          negative regulation of cAMP catabolic process
          negative regulation of heterotypic cell-cell adhesion
          positive regulation of transcription from RNA polymerase II promoter in response to stress
          negative regulation of MAP kinase activity
          regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          positive regulation of protein metabolic process
          negative regulation of cyclic-nucleotide phosphodiesterase activity
          negative regulation of NFAT protein import into nucleus
          negative regulation of response to cytokine stimulus
          cellular response to hydrogen peroxide
          negative regulation of ERK5 cascade
          cellular response to growth factor stimulus
          cellular response to laminar fluid shear stress
          cellular response to transforming growth factor beta stimulus
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of endothelial cell apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline