Search
Search

TNK1| Gene Symbol: TNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251902.1 | NP_001238831.1 | 13 | 2480 | ||
| NM_003985.4 | NP_003976.2 | 13 | 2465 | |||
| XM_011524045.2 | XP_011522347.1 | 13 | 2513 | |||
| XM_011524046.2 | XP_011522348.1 | 13 | 2596 | |||
| XM_011524047.2 | XP_011522349.1 | 12 | 2303 | |||
| XM_011524048.1 | XP_011522350.1 | 13 | 2370 | |||
| GenBank mRNA | AK225403.1 | - | 13 | 2348 | ||
| AK310684.1 | - | 14 | 2446 | |||
| BC035782.1 | - | 12 | 2330 | 5589312 | ||
| U43408.1 | - | 13 | 2365 | |||