Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL12B SIRNA Assays ›
          Gene Symbol:
          IL12B
          Gene Name:
          interleukin 12B
          Gene Aliases:
          CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
          Chromosome Location:
          Chr.5: 159314783 - 159330473 on Build GRCh38
          UniGene ID:
          Hs.674
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11089

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL12B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002187.2 NP_002178.2 3 251
          GenBank mRNA AF180563.1 - 2 215
          BC067498.1 - 2 213 6971868
          BC067499.1 - 2 213 6971869
          BC067500.1 - 2 213 6971872
          BC067502.1 - 2 213 6971874
          BC074723.2 - 3 240 30915173
          EF173865.1 - 2 177
          M65272.1 - 3 251
          M65290.1 - 2 222

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          natural killer cell activation involved in immune response
          positive regulation of T-helper 1 type immune response
          positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          negative regulation of inflammatory response to antigenic stimulus
          cell cycle arrest
          response to UV-B
          positive regulation of activation of JAK2 kinase activity
          cell migration
          cytokine-mediated signaling pathway
          sensory perception of pain
          sexual reproduction
          natural killer cell activation
          negative regulation of interleukin-10 production
          negative regulation of interleukin-17 production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell activation
          positive regulation of natural killer cell proliferation
          positive regulation of mononuclear cell proliferation
          positive regulation of tissue remodeling
          positive regulation of smooth muscle cell apoptotic process
          regulation of cytokine biosynthetic process
          T-helper 1 type immune response
          T-helper cell differentiation
          interferon-gamma biosynthetic process
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat4 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          defense response to protozoan
          positive regulation of memory T cell differentiation
          negative regulation of growth of symbiont in host
          positive regulation of interferon-gamma biosynthetic process
          positive regulation of osteoclast differentiation
          positive regulation of cell adhesion
          negative regulation of smooth muscle cell proliferation
          positive regulation of lymphocyte proliferation
          positive regulation of inflammatory response
          defense response to Gram-negative bacterium
          positive regulation of NK T cell activation
          positive regulation of NK T cell proliferation
          defense response to virus
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          cytokine receptor activity
          cytokine activity
          interleukin-12 receptor binding
          protein binding
          growth factor activity
          interleukin-12 alpha subunit binding
          identical protein binding
          protein homodimerization activity
          interleukin-23 receptor binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0