Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014796.1 | NP_001014796.1 | 4, 5 | 545 | ||
NM_006182.2 | NP_006173.2 | 3, 4 | 465 | |||
XM_006711344.3 | XP_006711407.1 | 3, 4 | 506 | |||
XM_011509586.2 | XP_011507888.1 | 3, 4 | 438 | |||
XM_011509587.2 | XP_011507889.1 | 4, 5 | 499 | |||
XM_011509588.2 | XP_011507890.1 | 3, 4 | 502 | |||
GenBank mRNA | AK095975.1 | - | 4, 5 | 518 | ||
AK225063.1 | - | 3, 4 | 384 | |||
AK225196.1 | - | 3, 4 | 440 | |||
AK314388.1 | - | 3, 4 | 414 | |||
BC052998.2 | - | 4, 5 | 516 | 6162716 | ||
BX640899.1 | - | 4, 5 | 502 | |||
CF994640.1 | - | 3, 4 | 515 | |||
X74764.1 | - | 3, 4 | 433 |