Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRMT2 SIRNA Assays ›
Gene Symbol:
PRMT2
Gene Name:
protein arginine methyltransferase 2
Gene Aliases:
HRMT1L1
Chromosome Location:
Chr.21: 46635153 - 46665685 on Build GRCh38
UniGene ID:
Hs.154163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110961
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRMT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242864.2
NP_001229793.1
4
491
NM_001242865.2
NP_001229794.1
4
491
NM_001242866.2
NP_001229795.1
4
491
NM_001286676.1
NP_001273605.1
4
491
NM_001286677.1
NP_001273606.1
4
491
NM_001286678.1
NP_001273607.1
4
491
NM_001535.4
NP_001526.2
4
491
NM_206962.3
NP_996845.1
5
600
XM_005261111.3
XP_005261168.1
5
1037
XM_006723998.3
XP_006724061.1
5
600
XM_006723999.2
XP_006724062.1
5
600
XM_006724000.2
XP_006724063.1
5
600
XM_017028339.1
XP_016883828.1
5
600
XM_017028340.1
XP_016883829.1
4
933
GenBank mRNA
AF059319.1
-
2
228
AK123352.1
-
4
1220
AK123650.1
-
5
1028
AK223157.1
-
4
412
AK312573.1
-
4
428
AL833586.1
-
4
418
AY786414.2
-
4
436
BC000727.2
-
5
520
3506339
BC038765.1
-
2
479
BC100026.1
-
4
405
30559810
BF683143.1
-
4
472
BQ421001.1
-
5
505
CR541804.1
-
3
266
FJ436410.1
-
4
442
FJ436411.1
-
4
442
FJ436412.1
-
4
442
KU178127.1
-
3
266
KU178128.1
-
3
266
U79286.1
-
4
433
U80213.1
-
4
431
X99209.1
-
4
442
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
protein methylation
signal transduction
histone methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
negative regulation of NF-kappaB transcription factor activity
histone arginine methylation
positive regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
developmental cell growth
regulation of androgen receptor signaling pathway
negative regulation of G1/S transition of mitotic cell cycle
transcription coactivator activity
signal transducer activity
protein binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
estrogen receptor binding
progesterone receptor binding
protein-arginine omega-N asymmetric methyltransferase activity
histone methyltransferase activity
protein homodimerization activity
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
thyroid hormone receptor binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0