Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 2 | 208 | ||
GenBank mRNA | AF294429.1 | - | 1 | 100 | ||
AK294921.1 | - | 2 | 207 | |||
AK304614.1 | - | 2 | 210 | |||
AK308578.1 | - | 2 | 241 | |||
AK308674.1 | - | 2 | 267 | |||
AK308879.1 | - | 2 | 237 | |||
AK309215.1 | - | 2 | 237 | |||
AK310427.1 | - | 2 | 190 | |||
AK311485.1 | - | 2 | 210 | |||
BC015942.1 | - | 2 | 208 | |||
BC034633.1 | - | 2 | 162 | |||
BC065732.1 | - | 2 | 189 | |||
BC136261.1 | - | 2 | 156 | 9020248 | ||
BC143547.1 | - | 2 | 156 | 9052058 | ||
BC151233.1 | - | 2 | 243 | |||
CR749504.1 | - | 2 | 184 |