Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP5 SIRNA Assays ›
          Gene Symbol:
          LRP5
          Gene Name:
          LDL receptor related protein 5
          Gene Aliases:
          BMND1, EVR1, EVR4, HBM, LR3, LRP-5, LRP7, OPPG, OPS, OPTA1, VBCH2
          Chromosome Location:
          Chr.11: 68298866 - 68449275 on Build GRCh38
          UniGene ID:
          Hs.6347
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111147

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRP5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291902.1 NP_001278831.1 2 477
          NM_002335.3 NP_002326.2 2 477
          XM_005273994.2 XP_005274051.1 2 472
          XM_011545029.1 XP_011543331.1 2 413
          XM_011545030.1 XP_011543332.1 2 413
          XM_011545031.1 XP_011543333.1 2 413
          GenBank mRNA AB017498.1 - 2 474
          AF064548.1 - 2 419
          AF077820.1 - 2 459
          AF088027.1 - 1 171
          BC098397.1 - 2 402
          BC150595.1 - 2 419 9056965
          DN999254.1 - 2 484

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          gastrulation with mouth forming second
          positive regulation of mesenchymal cell proliferation
          osteoblast development
          glucose catabolic process
          endocytosis
          cholesterol metabolic process
          regulation of blood pressure
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          Wnt signaling pathway
          positive regulation of osteoblast proliferation
          somatic stem cell population maintenance
          extracellular matrix-cell signaling
          cell migration involved in gastrulation
          cholesterol homeostasis
          embryonic digit morphogenesis
          regulation of apoptotic process
          Wnt signaling pathway involved in dorsal/ventral axis specification
          positive regulation of fat cell differentiation
          negative regulation of osteoblast differentiation
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          bone remodeling
          regulation of bone remodeling
          bone marrow development
          positive regulation of sequence-specific DNA binding transcription factor activity
          anatomical structure regression
          retina morphogenesis in camera-type eye
          canonical Wnt signaling pathway
          bone morphogenesis
          branching involved in mammary gland duct morphogenesis
          adipose tissue development
          cell-cell signaling involved in mammary gland development
          regulation of canonical Wnt signaling pathway
          regulation of insulin secretion involved in cellular response to glucose stimulus
          retinal blood vessel morphogenesis
          negative regulation of protein serine/threonine kinase activity
          apoptotic process involved in patterning of blood vessels
          beta-catenin destruction complex disassembly
          protein binding
          Wnt-protein binding
          toxin transporter activity
          Wnt-activated receptor activity
          coreceptor activity involved in Wnt signaling pathway
          coreceptor activity involved in canonical Wnt signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0