Search
Search

PNMT| Gene Symbol: PNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002686.4 | NP_002677.1 | 3 | 814 | ||
| NR_073461.1 | 3 | 679 | ||||
| XM_011524909.2 | XP_011523211.1 | 3 | 922 | |||
| GenBank mRNA | BC037246.1 | - | 3 | 1013 | 5175571 | |
| CR541815.1 | - | 3 | 791 | |||
| J03727.1 | - | 3 | 797 | |||