Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007371.3 | NP_031397.1 | 2, 3 | 403 | ||
XM_006717291.2 | XP_006717354.1 | 2, 3 | 535 | |||
XM_011519052.2 | XP_011517354.1 | 2, 3 | 754 | |||
XM_017015165.1 | XP_016870654.1 | 1, 2 | 328 | |||
GenBank mRNA | AK299519.1 | - | 2, 3 | 535 | ||
AY513270.1 | - | 1, 2 | 233 | |||
BC032124.2 | - | 2, 3 | 513 | 4856840 | ||
BI545861.1 | - | 2, 3 | 411 | |||
D26362.1 | - | 2, 3 | 354 | |||
KR633047.1 | - | 1, 2 | 215 |