siRNA Details
Gene Symbol: TOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003286.2 | NP_003277.1 | 12 | 1394 | ||
GenBank mRNA | AK292943.1 | - | 12 | 1393 | ||
BC136297.1 | - | 12 | 1245 | 9020284 | ||
J03250.1 | - | 12 | 1359 | |||
U07804.1 | - | 12 | 1138 | |||
U07806.1 | - | 12 | 1138 |
