Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111451
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
5
998
NM_001287743.1
NP_001274672.1
4
800
NM_001287744.1
NP_001274673.1
3
627
NM_001287745.1
NP_001274674.1
4
706
NM_001287747.1
NP_001274676.1
3
663
NM_001287748.1
NP_001274677.1
3
325
NM_001287749.1
NP_001274678.1
3
563
NM_001287750.1
NP_001274679.1
2
565
NM_001287751.1
NP_001274680.1
2
437
NM_004462.4
NP_004453.3
3
509
XM_017013217.1
XP_016868706.1
5
1290
GenBank mRNA
AK057726.1
-
3
627
AK098682.1
-
1083
AK293545.1
-
3
563
AK296043.1
-
3
381
AK297868.1
-
2
508
AK300059.1
-
2
327
AK300245.1
-
3
286
AK311246.1
-
2
276
AK311362.1
-
3
695
AK315993.1
-
4
576
AK316033.1
-
3
530
AK316182.1
-
2
525
AK316351.1
-
3
290
AK316531.1
-
3
275
AK316534.1
-
2
437
BC003573.1
-
3
356
3538137
BC009251.2
-
3
356
2961623
BC029641.1
-
3
319
4514761
BC034440.1
-
3
319
BG752446.1
-
373
BI765996.1
-
5
602
BT006704.1
-
3
287
CR457033.1
-
3
287
DC314061.1
-
3
485
DC387127.1
-
2
559
DC387479.1
-
3
325
DC396859.1
-
3
511
L06070.1
-
3
296
L06105.1
-
3
331
S76822.1
-
3
352
X69141.1
-
3
378
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00189506_m1
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0