Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG3 SIRNA Assays ›
          Gene Symbol:
          ALG3
          Gene Name:
          ALG3, alpha-1,3- mannosyltransferase
          Gene Aliases:
          CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
          Chromosome Location:
          Chr.3: 184242329 - 184249525 on Build GRCh38
          UniGene ID:
          Hs.478481
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111603
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006941.2 NP_001006942.1 9 1413
          NM_005787.5 NP_005778.1 9 1340
          NR_024533.1 8 1240
          NR_024534.1 9 1303
          XM_011512322.1 XP_011510624.1 9 1225
          XM_011512323.2 XP_011510625.1 9 1211
          GenBank mRNA AK289361.1 - 9 1369
          AK299575.1 - 8 1231
          BC002839.2 - 9 1315 3636604
          BC004313.1 - 9 1295
          BC021062.1 - 9 1304
          KP189279.1 - 9 1333
          KP189280.1 - 9 1296
          KP189281.1 - 8 1149
          KP189282.1 - 9 1450
          KP189283.1 - 8 1233
          KP189284.1 - 9 1331
          KP189285.1 - 9 1294
          KP189286.1 - 8 1149
          KP189287.1 - 9 1249
          KP189288.1 - 8 1231
          KP189289.1 - 9 1330
          KP189290.1 - 9 1294
          KP189291.1 - 8 1149
          KP189292.1 - 9 1249
          KP189293.1 - 8 1231
          KP189294.1 - 9 1309
          KP189295.1 - 8 1103
          KP189296.1 - 9 1272
          KP189297.1 - 8 1066
          KP189298.1 - 6 862
          KP189299.1 - 9 1165
          KP189300.1 - 8 1209
          KP189301.1 - 5 723
          KP189302.1 - 9 1330
          KP189303.1 - 9 1292
          KP189304.1 - 8 1149
          KP189305.1 - 9 1249
          KP189306.1 - 8 1229
          KP189307.1 - 9 1329
          KP189308.1 - 9 1292
          KP189309.1 - 8 1149
          KP189310.1 - 9 1249
          KP189311.1 - 8 1229
          KP189312.1 - 9 1329
          KP189313.1 - 8 1123
          KP189314.1 - 5 780
          KP189315.1 - 9 1292
          KP189316.1 - 8 1086
          KP189317.1 - 5 743
          KP189318.1 - 8 1149
          KP189319.1 - 7 943
          KP189320.1 - 4 600
          KP189321.1 - 9 1249
          KP189322.1 - 8 1043
          KP189323.1 - 4 539
          KP189324.1 - 8 1229
          KP189325.1 - 7 1023
          KP189326.1 - 4 680
          KP189327.1 - 3 519
          Y09022.1 - 9 1340

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00271780_m1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          dolichol-linked oligosaccharide biosynthetic process
          mannosylation
          alpha-1,3-mannosyltransferase activity
          dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-729pb:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline