Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B3GAT3 SIRNA Assays ›
          Gene Symbol:
          B3GAT3 GANAB
          Gene Name:
          beta-1,3-glucuronyltransferase 3
          glucosidase II alpha subunit
          Gene Aliases:
          GLCATI, JDSCD, glcUAT-I
          G2AN, GIIA, GLUII
          Chromosome Location:
          Chr.11: 62615296 - 62622175 on Build GRCh38
          Chromosome Location:
          Chr.11: 62624826 - 62646726 on Build GRCh38
          UniGene ID:
          Hs.502759
          Hs.595071
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 111785

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B3GAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288721.1 NP_001275650.1 3 691
          NM_001288722.1 NP_001275651.1 2 452
          NM_001288723.1 NP_001275652.1 2 452
          NM_012200.3 NP_036332.2 2 452
          NR_109991.1 2 442
          GenBank mRNA AB009598.1 - 2 253
          AJ005865.1 - 2 200
          AK075166.1 - 2 431
          AK297970.1 - 2 261
          AK310200.1 - 3 500
          AK316228.1 - 2 261
          BC007906.2 - 2 252 4299539
          BC019832.1 - 2 244
          BC071961.1 - 2 249 6214005
          Gene Symbol: GANAB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC005405.1 - 254

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001128184, BC166510, BC168710.
          This siRNA also targets the following transcripts in Mouse: NM_024256, AK079723, AK140424, AK048428, BC004038, and others.

          Panther Classification:

          Molecular Function -

          glycosyltransferase glucosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glycosaminoglycan biosynthetic process
          protein glycosylation
          heparan sulfate proteoglycan biosynthetic process
          glycosaminoglycan metabolic process
          chondroitin sulfate metabolic process
          positive regulation of catalytic activity
          chondroitin sulfate proteoglycan biosynthetic process
          dermatan sulfate proteoglycan biosynthetic process
          positive regulation of intracellular protein transport
          protein binding
          galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase activity
          glucuronosyltransferase activity
          metal ion binding
          protein phosphatase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0