Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MECR SIRNA Assays ›
Gene Symbol:
MECR
Gene Name:
mitochondrial trans-2-enoyl-CoA reductase
Gene Aliases:
CGI-63, FASN2B, NRBF1
Chromosome Location:
Chr.1: 29192657 - 29230958 on Build GRCh38
UniGene ID:
Hs.183646
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111938
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MECR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024732.2
NP_001019903.2
10
1291
NM_016011.3
NP_057095.3
10
1164
XM_005245885.2
XP_005245942.1
11
1214
XM_005245887.2
XP_005245944.1
12
1460
XM_011541539.1
XP_011539841.1
12
1388
XM_011541540.1
XP_011539842.1
11
1327
XM_011541541.1
XP_011539843.1
11
1243
XM_011541543.1
XP_011539845.1
12
1497
XM_011541545.1
XP_011539847.1
13
1661
XM_011541546.1
XP_011539848.1
11
1450
XM_011541547.1
XP_011539849.1
14
1783
XM_011541549.1
XP_011539851.1
13
1678
XM_011541550.1
XP_011539852.1
13
1533
XM_011541553.1
XP_011539855.1
13
1534
XM_011541554.1
XP_011539856.1
12
1298
XM_017001411.1
XP_016856900.1
13
1740
XM_017001412.1
XP_016856901.1
13
1655
XM_017001413.1
XP_016856902.1
12
1235
XM_017001414.1
XP_016856903.1
13
1482
XM_017001415.1
XP_016856904.1
12
1353
XM_017001416.1
XP_016856905.1
12
1389
GenBank mRNA
AF151821.1
-
10
1138
AK025830.1
-
10
1130
AK095099.1
-
10
1257
AK225687.1
-
10
1130
AK315274.1
-
10
1151
BC001419.2
-
10
1135
3139846
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025297, AK002317, AK002533, BC003864, BY693002, and others.
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
fatty acid biosynthetic process
oxidation-reduction process
zinc ion binding
trans-2-enoyl-CoA reductase (NADPH) activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0