siRNA Details
Gene Symbol: TLR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016610.3 | NP_057694.2 | 3 | 724 | ||
NM_138636.5 | NP_619542.1 | 2 | 587 | |||
XM_011545529.1 | XP_011543831.1 | 3 | 875 | |||
XM_011545530.2 | XP_011543832.1 | 3 | 703 | |||
GenBank mRNA | AB445666.1 | - | 1 | 500 | ||
AF245703.1 | - | 2 | 548 | |||
AF246971.1 | - | 3 | 705 | |||
AK314240.1 | - | 2 | 588 | |||
AY358296.1 | - | 2 | 556 | |||
BC101074.1 | - | 2 | 554 | 40008787 | ||
BC101075.2 | - | 2 | 554 | 40008788 | ||
BC101076.2 | - | 2 | 554 | 40008792 | ||
BC101077.1 | - | 2 | 554 | 40008793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
