Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR9 SIRNA Assays ›
          Gene Symbol:
          TLR9
          Gene Name:
          toll like receptor 9
          Gene Aliases:
          CD289
          Chromosome Location:
          Chr.3: 52221080 - 52226163 on Build GRCh38
          UniGene ID:
          Hs.87968
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112000

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLR9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017442.3 NP_059138.1 2 1662
          GenBank mRNA AB045180.1 - 2 1134
          AB445673.1 - 1 1028
          AF245704.1 - 2 1172
          AF259262.1 - 2 1662
          AF259263.1 - 1 904
          AY359085.1 - 1 1038
          BC032713.1 - 2 1124 5495717

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          response to molecule of bacterial origin
          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          I-kappaB phosphorylation
          axonogenesis
          positive regulation of gene expression
          maintenance of gastrointestinal epithelium
          negative regulation of NF-kappaB transcription factor activity
          tumor necrosis factor production
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          positive regulation of chemokine production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-18 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          negative regulation of toll-like receptor signaling pathway
          positive regulation of toll-like receptor signaling pathway
          toll-like receptor 9 signaling pathway
          phosphatidylinositol-3-phosphate biosynthetic process
          positive regulation of NF-kappaB import into nucleus
          defense response to bacterium
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JUN kinase activity
          positive regulation of interferon-gamma biosynthetic process
          innate immune response
          positive regulation of interferon-alpha biosynthetic process
          positive regulation of interferon-beta biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          regulation of cytokine secretion
          positive regulation of inflammatory response
          defense response to Gram-negative bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          negative regulation of calcium-transporting ATPase activity
          transmembrane signaling receptor activity
          interleukin-1 receptor binding
          1-phosphatidylinositol-3-kinase activity
          siRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0