Search
Search

UGT2B4| Gene Symbol: UGT2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297615.1 | NP_001284544.1 | 1 | 258 | ||
| NM_021139.2 | NP_066962.2 | 1 | 258 | |||
| GenBank mRNA | AF064200.1 | - | 1 | 236 | ||
| AF081793.1 | - | 1 | 246 | |||
| AJ005162.1 | - | 1 | 244 | |||
| AK292748.1 | - | 1 | 258 | |||
| AY529122.1 | - | 1 | 211 | |||
| BC026264.1 | - | 1 | 255 | 4709436 | ||
| BX494777.1 | - | 1 | 258 | |||
| DB185148.1 | - | 1 | 258 | |||
| Y00317.1 | - | 1 | 248 | |||