Search
Search

KRT19| Gene Symbol: KRT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002276.4 | NP_002267.2 | 3, 4 | 810 | ||
| GenBank mRNA | AB041267.1 | - | 2, 3 | 238 | ||
| AB041268.1 | - | 2, 3 | 238 | |||
| AK313261.1 | - | 3, 4 | 729 | |||
| BC002539.2 | - | 3, 4 | 714 | 3138504 | ||
| BC007628.1 | - | 3, 4 | 714 | 3504206 | ||
| BC010409.2 | - | 3, 4 | 721 | 3689178 | ||
| BC067744.1 | - | 3, 4 | 729 | 30332499 | ||
| BC084574.1 | - | 3, 4 | 712 | 6730698 | ||
| Y00503.1 | - | 3, 4 | 700 | |||