Search
Search

KRT85| Gene Symbol: KRT85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300810.1 | NP_001287739.1 | 5, 6 | 864 | ||
| NM_002283.3 | NP_002274.1 | 7, 8 | 1418 | |||
| GenBank mRNA | AK302303.1 | - | 5, 6 | 864 | ||
| X99140.1 | - | 7, 8 | 1364 | |||