Search
Search

CHST14| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130468.3 | NP_569735.1 | 1 | 1629 | ||
| GenBank mRNA | AB066595.1 | - | 1 | 1592 | ||
| AF282905.1 | - | 1 | 1428 | |||
| AF401222.1 | - | 1 | 1376 | |||
| AK074739.1 | - | 1 | 1384 | |||
| AY358446.1 | - | 1 | 1273 | |||
| BC023653.2 | - | 1 | 1263 | |||
| BC049214.1 | - | 1 | 1576 | 5518321 | ||
| BC053633.1 | - | 1 | 1535 | 6501263 | ||