Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001056.3 | NP_001047.1 | 8 | 1521 | ||
NM_176825.2 | NP_789795.1 | 9 | 1554 | |||
XM_005264013.3 | XP_005264070.1 | 8 | 1342 | |||
GenBank mRNA | AB008164.1 | - | 8 | 1191 | ||
AF186251.1 | - | 8 | 1386 | |||
AF186252.1 | - | 8 | 1521 | |||
AF186253.1 | - | 8 | 1237 | |||
AF186254.1 | - | 9 | 1419 | |||
AF186255.1 | - | 9 | 1554 | |||
AF186256.1 | - | 9 | 1270 | |||
AK297086.1 | - | 8 | 1327 | |||
AL832145.1 | - | 4 | 6295 | |||
BC005353.1 | - | 8 | 1138 | 4051264 | ||
U66036.1 | - | 8 | 1244 |