Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 8 | 914 | |||
NR_003227.1 | 7 | 741 | ||||
NR_003228.1 | 8 | 880 | ||||
GenBank mRNA | AF329691.1 | - | 7 | 665 | ||
AF329692.1 | - | 2 | 216 | |||
AK056488.1 | - | 8 | 885 | |||
AK091293.1 | - | 8 | 877 | |||
AK097044.1 | - | 7 | 728 | |||
AK126911.1 | - | 6 | 705 | |||
AK128352.1 | - | 1 | 580 | |||
AL832984.1 | - | 2 | 138 | |||
BC021245.2 | - | 6 | 487 |