Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAPDH SIRNA Assays ›
          Gene Symbol:
          GAPDH
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          Gene Aliases:
          G3PD, GAPD, HEL-S-162eP
          Chromosome Location:
          Chr.12: 6534405 - 6538375 on Build GRCh38
          UniGene ID:
          Hs.544577
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112842

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAPDH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256799.2 NP_001243728.1 7 1142
          NM_001289745.1 NP_001276674.1 8 1200
          NM_001289746.1 NP_001276675.1 7 1094
          NM_002046.5 NP_002037.2 8 1108
          GenBank mRNA AB062273.1 - 8 972
          AF261085.1 - 8 1023
          AK026525.1 - 8 1045
          AK308198.1 - 8 996
          AY007133.1 - 8 972
          AY633612.1 - 7 944
          BC001601.1 - 8 974 3543589
          BC004109.2 - 8 972 3678259
          BC009081.1 - 8 981 3869809
          BC013310.2 - 8 980 3538383
          BC020308.1 - 7 958
          BC023632.2 - 7 944 4842418
          BC025925.1 - 8 977 2989348
          BC026907.1 - 7 958 4454083
          BC029340.1 - 8 966
          BC029618.1 - 8 974 3876535
          BC083511.1 - 8 972 3536969
          BT006893.1 - 7 920
          CR407671.1 - 7 920
          EU668321.1 - 8 1022
          J02642.1 - 7 921
          M17851.1 - 7 953
          M28283.1 - 1 47
          M33197.1 - 8 980
          X01677.1 - 7 980
          X53778.1 - 8 973

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          gluconeogenesis
          regulation of macroautophagy
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          canonical glycolysis
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          protein binding
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding
          NADP binding
          NAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wksrx:80/100.66.72.56:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0