Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other P2RX7 SIRNA Assays ›
          Gene Symbol:
          P2RX7
          Gene Name:
          purinergic receptor P2X 7
          Gene Aliases:
          P2X7
          Chromosome Location:
          Chr.12: 121132819 - 121189478 on Build GRCh38
          UniGene ID:
          Hs.729169
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112943

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: P2RX7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002562.5 NP_002553.3 11, 12 1336
          NR_033948.1 11, 12 1559
          NR_033949.1 12, 13 1475
          NR_033950.1 9, 10 1436
          NR_033951.1 10, 11 1420
          NR_033952.1 9, 10 1347
          NR_033953.1 9, 10 1260
          NR_033954.1 10, 11 1239
          NR_033955.1 10, 11 1199
          NR_033956.1 9, 10 1126
          XM_011538419.2 XP_011536721.1 10, 11 1176
          XM_011538420.2 XP_011536722.1 9, 10 843
          XM_017019364.1 XP_016874853.1 11, 12 1489
          XM_017019365.1 XP_016874854.1 10, 11 1217
          XM_017019366.1 XP_016874855.1 10, 11 1204
          XM_017019367.1 XP_016874856.1 9, 10 1042
          GenBank mRNA AB209709.1 - 10, 11 1341
          AK090866.1 - 9, 10 1137
          AK225163.1 - 11, 12 1222
          AK290405.1 - 11, 12 1284
          AK296432.1 - 10, 11 1368
          AY847298.1 - 10, 11 1277
          AY847299.1 - 9, 10 1204
          AY847300.1 - 10, 11 1096
          AY847301.1 - 8, 9 1010
          AY847302.1 - 9, 10 983
          AY847303.1 - 11, 12 1416
          AY847304.1 - 12, 13 1332
          BC007679.2 - 10, 11 1137 3628076
          BC011913.2 - 11, 12 1262 4298811
          BC121157.1 - 11, 12 1262
          BC121158.1 - 9, 10 1362
          DQ399293.1 - 10, 11 1056
          GQ180122.1 - 11, 12 1202
          JX471562.1 - 1, 2 141
          Y09561.1 - 11, 12 1219

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell morphogenesis
          phagolysosome assembly
          positive regulation of T cell mediated cytotoxicity
          regulation of sodium ion transport
          protein phosphorylation
          membrane protein ectodomain proteolysis
          phospholipid transfer to membrane
          membrane budding
          inflammatory response
          mitochondrion organization
          cell surface receptor signaling pathway
          blood coagulation
          response to mechanical stimulus
          response to zinc ion
          positive regulation of calcium ion transport into cytosol
          positive regulation of gene expression
          positive regulation of glutamate secretion
          positive regulation of gamma-aminobutyric acid secretion
          synaptic vesicle exocytosis
          protein processing
          phospholipid scrambling
          sensory perception of pain
          cytolysis
          positive regulation of bone mineralization
          cellular response to extracellular stimulus
          bleb assembly
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          positive regulation of interleukin-6 production
          collagen metabolic process
          response to ATP
          response to fluid shear stress
          positive regulation of ion transmembrane transport
          purinergic nucleotide receptor signaling pathway
          T cell proliferation
          response to drug
          T cell homeostasis
          NAD transport
          negative regulation of MAPK cascade
          multicellular organismal protein catabolic process
          phospholipid translocation
          negative regulation of bone resorption
          negative regulation of cell volume
          positive regulation of glycolytic process
          positive regulation of cytolysis
          ceramide biosynthetic process
          pore complex assembly
          skeletal system morphogenesis
          homeostasis of number of cells within a tissue
          positive regulation of interleukin-1 alpha secretion
          positive regulation of interleukin-1 beta secretion
          defense response to Gram-positive bacterium
          release of sequestered calcium ion into cytosol
          protein oligomerization
          positive regulation of cytoskeleton organization
          response to calcium ion
          response to electrical stimulus
          regulation of killing of cells of other organism
          membrane depolarization
          positive regulation of mitochondrial depolarization
          positive regulation of lymphocyte apoptotic process
          cellular response to dsRNA
          reactive oxygen species metabolic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          positive regulation of bleb assembly
          lipopolysaccharide binding
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          receptor binding
          protein binding
          ATP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0