Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077500.1 | NP_001070968.1 | 4 | 526 | ||
NM_002776.4 | NP_002767.2 | 4 | 625 | |||
NM_145888.2 | NP_665895.1 | 4 | 487 | |||
XM_005259061.3 | XP_005259118.1 | 4 | 547 | |||
XM_005259062.3 | XP_005259119.1 | 4 | 511 | |||
XM_006723287.3 | XP_006723350.1 | 4 | 514 | |||
XM_006723289.3 | XP_006723352.1 | 4 | 609 | |||
XM_017026993.1 | XP_016882482.1 | 4 | 524 | |||
GenBank mRNA | AF024605.1 | - | 4 | 487 | ||
AK292365.1 | - | 4 | 616 | |||
AK309456.1 | - | 3 | 713 | |||
AY561635.1 | - | 4 | 554 | |||
BC002710.2 | - | 4 | 625 | 3632557 | ||
BM830577.1 | - | 4 | 526 | |||
CR450327.1 | - | 3 | 406 | |||
KJ010190.1 | - | 3 | 339 | |||
KJ010191.1 | - | 3 | 339 | |||
KJ010196.1 | - | 1 | 158 | |||
KJ010197.1 | - | 3 | 339 | |||
KJ010198.1 | - | 2 | 158 | |||
KJ010199.1 | - | 1 | 160 | |||
KJ010200.1 | - | 1 | 159 | |||
KJ010201.1 | - | 2 | 340 | |||
KJ010202.1 | - | 2 | 158 |