Search
Search

ADAM12| Gene Symbol: ADAM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288973.1 | NP_001275902.1 | 8, 9 | 1153 | ||
| NM_001288974.1 | NP_001275903.1 | 8, 9 | 1153 | |||
| NM_001288975.1 | NP_001275904.1 | 8, 9 | 1153 | |||
| NM_003474.5 | NP_003465.3 | 8, 9 | 1162 | |||
| NM_021641.4 | NP_067673.2 | 8, 9 | 1162 | |||
| XM_017016705.1 | XP_016872194.1 | 7, 8 | 1065 | |||
| GenBank mRNA | AF023476.2 | - | 8, 9 | 1055 | ||
| AF023477.2 | - | 8, 9 | 1055 | |||
| AK127856.1 | - | 8, 9 | 1058 | |||
| AK291629.1 | - | 8, 9 | 972 | |||
| AY358878.1 | - | 8, 9 | 1050 | |||
| BC060804.1 | - | 8, 9 | 1092 | 30348514 | ||
| KF444157.1 | - | 8, 9 | 1153 | |||