Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013283.4 | NP_037415.1 | 2 | 314 | ||
NM_182796.2 | NP_877725.1 | 2 | 453 | |||
GenBank mRNA | AB073390.1 | - | 2 | 228 | ||
AF113225.1 | - | 2 | 110 | |||
AF182814.1 | - | 2 | 155 | |||
AJ243721.1 | - | 2 | 293 | |||
AK096178.1 | - | 2 | 216 | |||
AK291919.1 | - | 2 | 264 | |||
AK294069.1 | - | 2 | 126 | |||
AK312365.1 | - | 2 | 263 | |||
AL136664.1 | - | 2 | 227 | |||
AU279489.1 | - | 2 | 263 | |||
AW966728.1 | - | 2 | 292 | |||
AY358695.1 | - | 2 | 208 | |||
BC005218.1 | - | 2 | 234 | 3931953 | ||
BC066645.1 | - | 2 | 189 | 5749043 | ||
BC093030.1 | - | 2 | 223 | 30527606 | ||
DA146198.1 | - | 2 | 317 | |||
DQ395260.1 | - | 3 | 295 | |||
DQ413183.1 | - | 2 | 292 |