Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135703.2 | NP_001129175.1 | 2 | 234 | ||
NM_013437.4 | NP_038465.1 | 3 | 291 | |||
GenBank mRNA | AF064841.1 | - | 2 | 226 | ||
AF166350.1 | - | 3 | 206 | |||
AK289752.1 | - | 3 | 195 | |||
AK299374.1 | - | 2 | 231 | |||
AL698796.1 | - | 3 | 259 | |||
BC032109.2 | - | 3 | 167 | 4842797 | ||
CR749434.1 | - | 2 | 206 | |||
DC372924.1 | - | 2 | 231 |