Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@1345c0e4
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
UCHL5
Gene Name:
ubiquitin C-terminal hydrolase L5
Gene Aliases:
CGI-70, INO80R, UCH-L5, UCH37
Chromosome Location:
Chr.1: 193012366 - 193060140 on Build GRCh38
UniGene ID:
Hs.145469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
113500
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price (USD)
233.00
Your Price (USD)
Online offer (USD):
233.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCHL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199261.1
NP_001186190.1
4
415
NM_001199262.1
NP_001186191.1
4
415
NM_001199263.1
NP_001186192.1
4
415
NM_015984.3
NP_057068.1
4
415
NR_037607.1
4
681
XM_005245244.4
XP_005245301.1
4
498
XM_005245246.4
XP_005245303.1
4
498
XM_005245247.4
XP_005245304.1
4
350
XM_005245248.4
XP_005245305.1
4
373
XM_005245249.4
XP_005245306.1
4
269
XM_006711366.3
XP_006711429.1
4
498
XM_006711367.3
XP_006711430.1
4
498
XM_006711368.3
XP_006711431.1
4
498
XM_006711369.2
XP_006711432.1
4
498
XM_006711370.3
XP_006711433.3
4
451
XM_006711371.3
XP_006711434.1
4
269
XM_011509604.1
XP_011507906.1
4
498
XM_011509607.2
XP_011507909.1
4
309
XM_011509608.2
XP_011507910.1
5
670
XM_011509609.2
XP_011507911.1
5
574
XM_017001429.1
XP_016856918.1
4
498
XM_017001430.1
XP_016856919.1
4
498
XM_017001431.1
XP_016856920.1
4
498
XM_017001432.1
XP_016856921.1
4
498
XM_017001433.1
XP_016856922.1
4
498
XM_017001434.1
XP_016856923.1
4
451
XM_017001435.1
XP_016856924.1
4
451
XM_017001436.1
XP_016856925.1
5
670
XM_017001437.1
XP_016856926.1
5
670
XM_017001438.1
XP_016856927.1
4
293
XM_017001439.1
XP_016856928.1
4
714
XM_017001440.1
XP_016856929.1
4
678
XM_017001441.1
XP_016856930.1
4
654
XM_017001442.1
XP_016856931.1
4
269
XM_017001443.1
XP_016856932.1
4
269
XM_017001444.1
XP_016856933.1
4
663
GenBank mRNA
AF147717.1
-
4
413
AF151828.1
-
4
295
AF157320.1
-
4
430
AK024751.1
-
4
361
AK225794.1
-
4
348
AK301381.1
-
4
681
AK314445.1
-
4
359
BC015521.1
-
4
348
3912584
BC025369.2
-
4
393
3937407
BT006790.1
-
4
282
DA255130.1
-
4
415
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
hydrolase
protease
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-blue:8080/10.253.225.14:8080.
git-commit: 578676318d66c763e4a01d7f975a961cd389eaca
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.18.0-2021.03.09-1.0