Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1G SIRNA Assays ›
          Gene Symbol:
          CACNA1G
          Gene Name:
          calcium voltage-gated channel subunit alpha1 G
          Gene Aliases:
          Ca(V)T.1, Cav3.1, NBR13, SCA42
          Chromosome Location:
          Chr.17: 50560929 - 50629625 on Build GRCh38
          UniGene ID:
          Hs.591169
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 113584

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256324.1 NP_001243253.1 5 1117
          NM_001256325.1 NP_001243254.1 5 1117
          NM_001256326.1 NP_001243255.1 5 1117
          NM_001256327.1 NP_001243256.1 5 1117
          NM_001256328.1 NP_001243257.1 5 1117
          NM_001256329.1 NP_001243258.1 5 1117
          NM_001256330.1 NP_001243259.1 5 1117
          NM_001256331.1 NP_001243260.1 5 1117
          NM_001256332.1 NP_001243261.1 5 1117
          NM_001256333.1 NP_001243262.1 5 1117
          NM_001256334.1 NP_001243263.1 5 1117
          NM_001256359.1 NP_001243288.1 5 1117
          NM_001256360.1 NP_001243289.1 5 1117
          NM_001256361.1 NP_001243290.1 5 1117
          NM_018896.4 NP_061496.2 5 1117
          NM_198376.2 NP_938190.1 5 1117
          NM_198377.2 NP_938191.2 5 1117
          NM_198378.2 NP_938192.1 5 1117
          NM_198379.2 NP_938193.1 5 1117
          NM_198380.2 NP_938194.1 5 1117
          NM_198382.2 NP_938196.1 5 1117
          NM_198383.2 NP_938197.1 5 1117
          NM_198384.2 NP_938198.1 5 1117
          NM_198385.2 NP_938199.1 5 1117
          NM_198386.2 NP_938200.1 5 1117
          NM_198387.2 NP_938201.1 5 1117
          NM_198388.2 NP_938202.1 5 1117
          NM_198396.2 NP_938406.1 5 1117
          NR_046054.1 5 1117
          NR_046055.1 5 1117
          NR_046056.1 5 1117
          NR_046057.1 5 1117
          NR_046058.1 5 1117
          XM_006722160.3 XP_006722223.1 5 1256
          XM_006722161.3 XP_006722224.1 5 1256
          GenBank mRNA AB012043.1 - 5 753
          AF124351.1 - 5 1097
          AF126965.1 - 5 900
          AF126966.1 - 5 900
          AF134985.1 - 5 725
          AF134986.1 - 5 725
          AF190860.1 - 5 941
          AF227744.1 - 5 725
          AF227745.1 - 5 725
          AF227746.1 - 5 725
          AF227747.1 - 5 725
          AF227748.1 - 5 725
          AF227749.1 - 5 725
          AF227750.1 - 5 725
          AF227751.1 - 5 725
          BC110995.1 - 5 1117 5502027
          DQ494449.1 - 5 725
          DQ494450.1 - 5 725
          DQ494451.1 - 5 725
          DQ494452.1 - 5 725
          DQ494453.1 - 5 725
          DQ494454.1 - 5 725
          DQ494455.1 - 5 725
          DQ494456.1 - 5 725
          DQ494457.1 - 5 725
          DQ494458.1 - 5 725
          DQ494459.1 - 5 725
          DQ494460.1 - 5 725
          DQ494461.1 - 5 725
          DQ494462.1 - 5 725
          DQ494463.1 - 5 725
          DQ494464.1 - 5 725
          DQ494465.1 - 5 725
          DQ494466.1 - 5 725
          DQ494467.1 - 5 725
          DQ494468.1 - 5 725
          DQ494469.1 - 5 725
          DQ494470.1 - 5 725
          DQ494471.1 - 5 725
          DQ494472.1 - 5 725
          DQ494473.1 - 5 725
          DQ494474.1 - 5 725
          DQ494475.1 - 5 725
          DQ494476.1 - 5 725
          DQ494477.1 - 5 725
          DQ494478.1 - 5 725
          DQ494479.1 - 5 725
          KJ534993.1 - 5 725
          KJ535064.1 - 5 708

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          response to nickel cation
          regulation of ion transmembrane transport
          regulation of membrane potential
          positive regulation of calcium ion-dependent exocytosis
          regulation of atrial cardiac muscle cell membrane depolarization
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          SA node cell action potential
          AV node cell action potential
          SA node cell to atrial cardiac muscle cell signalling
          AV node cell to bundle of His cell signaling
          membrane depolarization during AV node cell action potential
          membrane depolarization during SA node cell action potential
          regulation of heart rate by cardiac conduction
          low voltage-gated calcium channel activity
          voltage-gated calcium channel activity involved in AV node cell action potential
          voltage-gated calcium channel activity involved SA node cell action potential
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0