Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MST1 SIRNA Assays ›
          Gene Symbol:
          MST1 MST1P2
          Gene Name:
          macrophage stimulating 1
          macrophage stimulating 1 pseudogene 2
          Gene Aliases:
          D3F15S2, DNF15S2, HGFL, MSP, NF15S2
          MSPL-1, MSPL-2, MSPL-3, MSPL1, MSPL2, MSPL3, MSTP1, MSTP2, MSTP3
          Chromosome Location:
          Chr.3: 49683947 - 49689528 on Build GRCh38
          Chromosome Location:
          Chr.1: 16645574 - 16650420 on Build GRCh38
          UniGene ID:
          Hs.349110
          Hs.349110
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 113635

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020998.3 NP_066278.3 3, 4 419
          XM_006713166.1 XP_006713229.1 3, 4 1184
          XM_011533732.1 XP_011532034.1 3, 4 1184
          XM_011533737.2 XP_011532039.1 3, 4 1184
          XM_011533738.2 XP_011532040.1 3, 4 1184
          XM_017006460.1 XP_016861949.1 3, 4 1184
          XM_017006461.1 XP_016861950.1 3, 4 1184
          XM_017006462.1 XP_016861951.1 3, 4 1184
          XM_017006463.1 XP_016861952.1 3, 4 1184
          XM_017006464.1 XP_016861953.1 3, 4 1184
          XM_017006465.1 XP_016861954.1 3, 4 1184
          XM_017006466.1 XP_016861955.1 3, 4 1184
          XM_017006467.1 XP_016861956.1 3, 4 1184
          XM_017006468.1 XP_016861957.1 3, 4 1184
          GenBank mRNA AK222893.1 - 3, 4 310
          AK294324.1 - 3, 4 419
          AK298396.1 - 3, 4 329
          AK298452.1 - 2, 3 179
          BC044862.1 - 2, 3 1867
          BC048330.1 - 3, 4 318 5190966
          DC315638.1 - 3, 4 419
          L11924.1 - 3, 4 315
          M74178.1 - 3, 4 305
          Gene Symbol: MST1P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK298417.1 - 3, 4 324

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          negative regulation of gluconeogenesis
          hepatocyte growth factor receptor signaling pathway
          regulation of cAMP-dependent protein kinase activity
          molecular_function
          serine-type endopeptidase activity
          protein binding
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0