Search
Search

USP46| Gene Symbol: USP46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134223.1 | NP_001127695.1 | 5 | 878 | ||
| NM_001286767.1 | NP_001273696.1 | 5 | 766 | |||
| NM_001286768.1 | NP_001273697.1 | 6 | 910 | |||
| NM_022832.3 | NP_073743.2 | 5 | 802 | |||
| XM_017008557.1 | XP_016864046.1 | 5 | 768 | |||
| GenBank mRNA | AK022614.1 | - | 5 | 622 | ||
| AK024318.1 | - | 5 | 743 | |||
| AK296493.1 | - | 6 | 883 | |||
| AK299883.1 | - | 5 | 679 | |||
| AK302438.1 | - | 5 | 877 | |||
| BC037574.1 | - | 5 | 751 | 5529667 | ||
| GU455414.1 | - | 5 | 617 | |||