Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002370.3 | NP_002361.1 | 2, 3 | 317 | ||
GenBank mRNA | AF035940.1 | - | 2, 3 | 220 | ||
AF067173.1 | - | 2, 3 | 243 | |||
AK312113.1 | - | 2, 3 | 253 | |||
BC018211.1 | - | 2, 3 | 229 | 3861094 | ||
BX283982.1 | - | 2, 3 | 317 |