Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183062.2 | NP_898885.1 | 5 | 990 | ||
GenBank mRNA | BC130400.1 | - | 5 | 1062 | 40146431 |