Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198185.4 | NP_937828.3 | 4 | 365 | ||
XM_017017659.1 | XP_016873148.1 | 4 | 865 |