Search
Search

CHRNB3| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000749.3 | NP_000740.1 | 5 | 1222 | ||
| XM_011544390.1 | XP_011542692.1 | 3 | 1045 | |||
| GenBank mRNA | BC069681.1 | - | 5 | 1222 | 7262491 | |
| BC069703.1 | - | 5 | 1222 | 7262503 | ||
| BC069788.1 | - | 5 | 1222 | 7262515 | ||
| U62438.1 | - | 5 | 1190 | |||
| X67513.1 | - | 4 | 989 | |||
| Y08417.1 | - | 5 | 1222 | |||