Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HOXB3 SIRNA Assays ›
          Gene Symbol:
          HOXB3
          Gene Name:
          homeobox B3
          Gene Aliases:
          HOX2, HOX2G, Hox-2.7
          Chromosome Location:
          Chr.17: 48548870 - 48590369 on Build GRCh38
          UniGene ID:
          Hs.654560
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 114211

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HOXB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002146.4 NP_002137.4 4 2368
          XM_005257277.3 XP_005257334.1 5 2082
          XM_005257280.3 XP_005257337.1 5 1993
          XM_005257282.3 XP_005257339.1 5 1558
          XM_006721854.2 XP_006721917.1 5 2100
          XM_011524707.2 XP_011523009.1 5 11908
          XM_011524708.2 XP_011523010.1 4 11488
          XM_011524710.1 XP_011523012.1 5 4308
          XM_011524719.1 XP_011523021.1 4 6789
          XM_011524720.2 XP_011523022.1 4 9480
          XM_011524721.2 XP_011523023.1 4 4612
          XM_011524726.2 XP_011523028.1 4 1566
          XM_017024557.1 XP_016880046.1 5 11656
          XM_017024558.1 XP_016880047.1 5 11530
          XM_017024559.1 XP_016880048.1 5 11372
          XM_017024560.1 XP_016880049.1 5 11616
          XM_017024561.1 XP_016880050.1 5 15453
          XM_017024562.1 XP_016880051.1 5 11394
          XM_017024563.1 XP_016880052.1 6 14049
          GenBank mRNA AK027609.1 - 4 2406
          AK291182.1 - 5 2079
          AK316245.1 - 4 1601
          U59298.1 - 2 2959
          X16667.1 - 4 1873

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          hematopoietic progenitor cell differentiation
          transcription, DNA-templated
          anterior/posterior pattern specification
          rhombomere development
          glossopharyngeal nerve morphogenesis
          thyroid gland development
          embryonic skeletal system morphogenesis
          regulation of neurogenesis
          cartilage development
          definitive hemopoiesis
          face development
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cdxgb:80/100.66.73.10:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0