Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003083.3 | NP_003074.1 | 5 | 828 | ||
NR_030717.1 | 5 | 725 | ||||
GenBank mRNA | AK222926.1 | - | 5 | 799 | ||
AK314878.1 | - | 5 | 813 | |||
BC011868.2 | - | 5 | 783 | 4545178 | ||
BX647416.1 | - | 5 | 692 | |||
U44755.1 | - | 5 | 812 | |||
U44898.1 | - | 5 | 794 |