Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003728.3 | NP_003719.3 | 3 | 729 | ||
XM_005263321.3 | XP_005263378.1 | 3 | 729 | |||
XM_017008789.1 | XP_016864278.1 | 4 | 7146 | |||
XM_017008790.1 | XP_016864279.1 | 3 | 374 | |||
GenBank mRNA | AF055634.1 | - | 3 | 530 | ||
AK290500.1 | - | 3 | 540 | |||
AK308421.1 | - | 3 | 492 | |||
BC041156.1 | - | 3 | 725 | 5208108 |