Search
Search

CHRNA6| Gene Symbol: CHRNA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199279.1 | NP_001186208.1 | 2 | 543 | ||
| NM_004198.3 | NP_004189.1 | 2 | 543 | |||
| GenBank mRNA | AK298798.1 | - | 2 | 367 | ||
| AK313521.1 | - | 2 | 389 | |||
| BC014456.1 | - | 2 | 233 | 4862198 | ||
| DC307404.1 | - | 2 | 545 | |||
| U62435.1 | - | 2 | 329 | |||
| Y16282.1 | - | 2 | 187 | |||