siRNA Details
Gene Symbol: POU3F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005604.3 | NP_005595.2 | 1 | 1673 | ||
GenBank mRNA | BC051699.1 | - | 1 | 1642 | 4817001 | |
Z11933.1 | - | 1 | 1665 |

Gene Symbol: POU3F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005604.3 | NP_005595.2 | 1 | 1673 | ||
GenBank mRNA | BC051699.1 | - | 1 | 1642 | 4817001 | |
Z11933.1 | - | 1 | 1665 |