Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC4 SIRNA Assays ›
          Gene Symbol:
          HDAC4
          Gene Name:
          histone deacetylase 4
          Gene Aliases:
          AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
          Chromosome Location:
          Chr.2: 239048168 - 239401647 on Build GRCh38
          UniGene ID:
          Hs.20516
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 114593

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006037.3 NP_006028.2 27 6344
          XM_006712877.3 XP_006712940.1 26 12290
          XM_006712878.3 XP_006712941.1 27 5864
          XM_006712879.3 XP_006712942.1 26 11757
          XM_006712880.3 XP_006712943.1 26 11769
          XM_011512217.2 XP_011510519.1 26 12305
          XM_011512218.2 XP_011510520.1 26 12290
          XM_011512219.2 XP_011510521.1 26 12275
          XM_011512220.2 XP_011510522.1 26 6044
          XM_011512221.1 XP_011510523.1 27 6032
          XM_011512222.2 XP_011510524.1 27 6333
          XM_011512223.2 XP_011510525.1 27 5847
          XM_011512224.2 XP_011510526.1 26 6235
          XM_011512225.2 XP_011510527.1 26 6220
          XM_011512226.2 XP_011510528.1 26 5792
          XM_011512227.2 XP_011510529.1 25 11319
          XM_011512230.1 XP_011510532.1 21 5230
          XM_017005394.1 XP_016860883.1 25 12174
          XM_017005395.1 XP_016860884.1 23 5096
          GenBank mRNA AB006626.2 - 27 6345

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast development
          chromatin remodeling
          transcription, DNA-templated
          inflammatory response
          nervous system development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of lamellipodium assembly
          negative regulation of myotube differentiation
          regulation of cardiac muscle contraction by calcium ion signaling
          response to denervation involved in regulation of muscle adaptation
          cardiac muscle hypertrophy in response to stress
          positive regulation of smooth muscle cell migration
          chromatin modification
          histone deacetylation
          B cell differentiation
          positive regulation of protein sumoylation
          peptidyl-lysine deacetylation
          regulation of gene expression, epigenetic
          B cell activation
          response to drug
          regulation of protein binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neuron apoptotic process
          negative regulation of osteoblast differentiation
          negative regulation of glycolytic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle cell proliferation
          regulation of skeletal muscle fiber development
          positive regulation of sequence-specific DNA binding transcription factor activity
          response to interleukin-1
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to mechanical stimulus
          cellular response to tumor necrosis factor
          cellular response to parathyroid hormone stimulus
          positive regulation of reactive oxygen species biosynthetic process
          RNA polymerase III transcription factor binding
          core promoter binding
          chromatin binding
          transcription corepressor activity
          histone deacetylase activity
          protein binding
          transcription factor binding
          zinc ion binding
          protein kinase binding
          potassium ion binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          activating transcription factor binding
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0