Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014213.3 | NP_055028.3 | 1, 2 | 896 | ||
GenBank mRNA | BC044855.1 | - | 1, 2 | 896 | 5191006 | |
X59372.1 | - | 1, 2 | 818 |