Search
Search

CD99| Gene Symbol: CD99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122898.2 | NP_001116370.1 | 9 | 1159 | ||
| NM_001321368.1 | NP_001308297.1 | 10 | 1204 | |||
| NM_001321369.1 | NP_001308298.1 | 9 | 1156 | |||
| NM_001321370.1 | NP_001308299.1 | 9 | 1156 | |||
| NM_002414.4 | NP_002405.1 | 10 | 1207 | |||
| NR_135623.1 | 11 | 1222 | ||||
| GenBank mRNA | BC002584.2 | - | 10 | 1085 | 3161673 | |
| BC003147.1 | - | 10 | 1043 | 2906143 | ||
| BC010109.2 | - | 10 | 1153 | 3606974 | ||
| BC021620.1 | - | 10 | 1043 | 2957883 | ||
| BC024310.1 | - | 10 | 1146 | 4094268 | ||
| M16279.1 | - | 10 | 1156 | |||