Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000816.3 | NP_000807.2 | 9 | 1525 | ||
NM_198903.2 | NP_944493.2 | 11 | 1669 | |||
NM_198904.2 | NP_944494.1 | 10 | 1549 | |||
GenBank mRNA | AK299640.1 | - | 10 | 1250 | ||
AK315200.1 | - | 10 | 1201 | |||
BC059389.1 | - | 10 | 1370 | 5288256 | ||
BC069348.1 | - | 9 | 1261 | 7262321 | ||
BC074795.2 | - | 9 | 1199 | 30915258 | ||
CD013896.1 | - | 10 | 1327 | |||
CD013897.1 | - | 9 | 1235 | |||
X15376.1 | - | 9 | 1393 |