Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRP2 SIRNA Assays ›
          Gene Symbol:
          NRP2
          Gene Name:
          neuropilin 2
          Gene Aliases:
          NP2, NPN2, PRO2714, VEGF165R2
          Chromosome Location:
          Chr.2: 205682450 - 205798133 on Build GRCh38
          UniGene ID:
          Hs.471200
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115160
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003872.2 NP_003863.2 2 994
          NM_018534.3 NP_061004.3 2 994
          NM_201264.1 NP_957716.1 2 994
          NM_201266.1 NP_957718.1 2 994
          NM_201267.1 NP_957719.1 2 994
          NM_201279.1 NP_958436.1 2 994
          XM_005246933.3 XP_005246990.2 3 859
          XM_005246934.3 XP_005246991.2 3 532
          XM_017005185.1 XP_016860674.1 3 858
          XM_017005186.1 XP_016860675.1 3 859
          XM_017005187.1 XP_016860676.1 2 1044
          XM_017005188.1 XP_016860677.1 2 1043
          GenBank mRNA AF016098.1 - 2 203
          AF022859.1 - 2 203
          AF022860.1 - 2 203
          AF280544.1 - 2 203
          AF280545.1 - 2 203
          AF280546.1 - 2 203
          AK130198.1 - 3 980
          AK290934.1 - 3 674
          AL833606.1 - 2 994
          BC009222.2 - 4 672
          BC101525.1 - 2 234 8069031
          BC104770.1 - 2 234 8143773
          BC117413.1 - 2 234 40125964
          BC143237.1 - 2 234
          BC143238.1 - 2 234 9051739
          BC143608.1 - 2 234 9052121
          BX537423.1 - 2 993
          KJ534899.1 - 2 203
          KJ534900.1 - 2 203

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_030869, XM_006245040, XM_006245039, XM_006245042, XM_006245041, and others.

          Matching Taqman Gene Expression Assay:


          Hs00187290_m1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of endothelial cell proliferation
          cell adhesion
          axon guidance
          positive regulation of endothelial cell migration
          nerve development
          vascular endothelial growth factor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          axon extension involved in axon guidance
          sympathetic ganglion development
          sympathetic neuron projection extension
          sympathetic neuron projection guidance
          semaphorin-plexin signaling pathway involved in neuron projection guidance
          receptor activity
          vascular endothelial growth factor-activated receptor activity
          heparin binding
          semaphorin receptor activity
          growth factor binding
          cytokine binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0