siRNA Details
Gene Symbol: SOX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000346.3 | NP_000337.1 | 3 | 3248 | ||
GenBank mRNA | BC018276.1 | - | 1 | 549 | ||
Z46629.1 | - | 3 | 3236 |

Gene Symbol: SOX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000346.3 | NP_000337.1 | 3 | 3248 | ||
GenBank mRNA | BC018276.1 | - | 1 | 549 | ||
Z46629.1 | - | 3 | 3236 |