Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040667.2 | NP_001035757.1 | 15 | 2385 | ||
NM_001538.3 | NP_001529.2 | 15 | 2295 | |||
GenBank mRNA | AB029348.1 | - | 13 | 1427 | ||
AK225152.1 | - | 11 | 1143 | |||
BC130383.1 | - | 13 | 1384 | |||
BG980713.1 | - | 1 | 97 | |||
BM768246.1 | - | 4 | 247 | |||
D87673.1 | - | 13 | 1337 |